Anti-GADD45B Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5904R-A350
10426-204EA 792.07 USD
10426-204
Anti-GADD45B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Involved in the regulation of growth and apoptosis. Mediates activation of stress-responsive MTK1/MEKK4 MAPKKK.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: GADD45B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR