Anti-BCL2L2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-5903R-A647
10426-186EA 792.07 USD
10426-186
Anti-BCL2L2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Promotes cell survival. Blocks dexamethasone-induced apoptosis. Mediates survival of postmitotic Sertoli cells by suppressing death-promoting activity of BAX.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR