Anti-OS9 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5901R-CY7
10425-828EA 792.07 USD
10425-828
Anti-OS9 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Lectin which functions in endoplasmic reticulum (ER) quality control and ER-associated degradation (ERAD). May bind terminally misfolded non-glycosylated proteins as well as improperly folded glycoproteins, retain them in the ER, and possibly transfer them to the ubiquitination machinery and promote their degradation. Possible targets include TRPV4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: OS9
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR