Anti-ADORA2B Rabbit Polyclonal Antibody (Biotin)

BS-5900R-BIO
10426-142EA 529.29 USD
10426-142
Anti-ADORA2B Rabbit Polyclonal Antibody (Biotin)
Receptor for adenosine. The activity of this receptor is mediated by G proteins which activate adenylyl cyclase.

Recommended Dilutions: Western Blot: 1:100-1000

Type: Primary
Antigen: ADORA2B
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR