Anti-HIF3A Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-5898R-CY5.5
10425-894EA 792.07 USD
10425-894
Anti-HIF3A Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Involved in adaptive response to hypoxia. Suppresses hypoxia-inducible expression of HIF1A and EPAS1. Binds to core DNA sequence 5'-TACGTG-3' within the hypoxia response element (HRE) of target gene promoters. The complex HIF3A-ARNT activates the transcription of reporter genes driven by HRE. Isoform 4 has a dominant-negative function of inactivating HIF1A-mediated transcription. Isoform 4 attenuates the binding of HIF1A to hypoxia-responsive elements (HRE), thus inhibiting HRE-driven transcription. Hypoxia induces down-regulation of isoform 4, leading to activation of HIF1A in hypoxia. Conversely, upon restoring normoxia, the expression of isoform 4 increases and thereby secure an inhibition of HIF1A activity. Isoform 4 may be a negative regulator of hypoxia-inducible gene expression in the kidney and may be involved in renal tumorigenesis. Functions as an inhibitor of angiogenesis in the cornea (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HIF3A
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR