Anti-EDN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss
BS-5897R-A555
10426-052EA 746.42 USD
10426-052
Anti-EDN3 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Endothelins are vasoactive proteins derived form the endothelium The active form of this protein is a 21aa peptide processed from the precursor. The active peptide binds endothelin receptor type B (EDNRB). This interaction EDNRB regulates the development of neural crest-derived cell lineages, such as melanocytes and enteric neurons. Mutations in Endothelin 3 and EDNRB have been associated with Hirschsprung disease (HSCR) and Waardenburg syndrome (WS), which are congenital disorders involving neural crest-derived cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EDN3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR