Anti-WFDC5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-5896R-A555
10426-028EA 792.07 USD
10426-028
Anti-WFDC5 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
WFDC5 is a member of the WAP-type four-disulfide core (WFDC) domain family. It is a putative acid-stable proteinase inhibitor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: WFDC5
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR