Anti-TBC1D4 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-5893R-CY5
10425-818EA 791.84 USD
10425-818
Anti-TBC1D4 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
May act as a GTPase-activating protein for RAB2A, RAB8A, RAB10 and RAB14. Isoform 2 promotes insulin-induced glucose transporter SLC2A4/GLUT4 translocation at the plasma membrane, thus increasing glucose uptake.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TBC1D4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR