Anti-LUM Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-5890R-A647
10425-972EA 792.07 USD
10425-972
Anti-LUM Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Present in the extracellular matrix of human articular cartilage at all ages, although its abundance is far greater in the adult. In the adult cartilage lumican exists predominantly in a glycoprotein form lacking keratan sulfate, whereas the juvenile form of the molecule is a proteoglycan.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LUM
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR