Anti-HYAL3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-5889R-CY5.5
10425-956EA 792.07 USD
10425-956
Anti-HYAL3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
HYAL3 is a protein which is similar in structure to hyaluronidases. Hyaluronidases intracellularly degrade hyaluronan, one of the major glycosaminoglycans of the extracellular matrix. Hyaluronan is thought to be involved in cell proliferation, migration and differentiation. However, this protein has not yet been shown to have hyaluronidase activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HYAL3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR