Anti-HYAL2 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5888R-CY3
10425-928EA 792.07 USD
10425-928
Anti-HYAL2 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Hydrolyzes high molecular weight hyaluronic acid to produce an intermediate-sized product which is further hydrolyzed by sperm hyaluronidase to give small oligosaccharides. Displays very low levels of activity. Associates with and negatively regulates MST1R.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HYAL2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR