Anti-MHC Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5885R-A350
10425-846EA 792.07 USD
10425-846
Anti-MHC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Myosin is a protein which is known to interact with actin in muscle and non-muscle cells. It contains two identical heavy chains and four light chains. Myosin molecules consist of two major regions: tails (rods) and heads. They aggregate into filaments through the tail region and interact with actin and with ATP through the head region. Multiple forms of myosin heavy chains exist for each muscle type-skeletal, cardiac, smooth and non-muscle isomyosin forms exist in different types of skeletal muscle, depending on the physiological function of the muscle. They are designated at type I (slow twitch) and type II (fast twitch).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MHC
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR