Anti-EMCN Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5884R-A350
10425-614EA 792.07 USD
10425-614
Anti-EMCN Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Endothelial sialomucin, also called endomucin or mucin-like sialoglycoprotein, which interferes with the assembly of focal adhesion complexes and inhibits interaction between cells and the extracellular matrix.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EMCN
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR