Anti-CTHRC1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5882R-CY7
10425-580EA 792.07 USD
10425-580
Anti-CTHRC1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
CTHRC1 is a protein that may play a role in the cellular response to arterial injury through involvement in vascular remodeling. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcript variants have been described.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTHRC1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR