Anti-CTHRC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-5882R-A647
10425-770EA 792.07 USD
10425-770
Anti-CTHRC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
CTHRC1 is a protein that may play a role in the cellular response to arterial injury through involvement in vascular remodeling. Mutations at this locus have been associated with Barrett esophagus and esophageal adenocarcinoma. Alternatively spliced transcript variants have been described.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: CTHRC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR