Anti-COL20A1 Rabbit Polyclonal Antibody (Biotin)

BS-5881R-BIO
10425-748EA 529.29 USD
10425-748
Anti-COL20A1 Rabbit Polyclonal Antibody (Biotin)
COL20A1 is a probable collagen protein.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: COL20A1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR