Anti-COL20A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-5881R-A488
10425-742EA 792.07 USD
10425-742
Anti-COL20A1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
COL20A1 is a probable collagen protein.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COL20A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR