Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5879R-A350
10425-690EA 792.07 USD
10425-690
Anti-MUC7 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May function in a protective capacity by promoting the clearance of bacteria in the oral cavity and aiding in mastication, speech, and swallowing. Binds P.aeruginosa pili.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MUC7
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR