Anti-MMP23B Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5875R-A350
10425-576EA 792.07 USD
10425-576
Anti-MMP23B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Protease. May regulate the surface expression of some potassium channels by retaining them in the endoplasmic reticulum (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MMP23B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR