Anti-KLK8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5871R-A350
10425-504EA 792.07 USD
10425-504
Anti-KLK8 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Serine protease which is capable of degrading a number of proteins such as casein, fibrinogen, kininogen, fibronectin and collagen type IV. Also cleaves L1CAM in response to increased neural activity. Induces neurite outgrowth and fasciculation of cultured hippocampal neurons. Plays a role in the formation and maturation of orphan and small synaptic boutons in the Schaffer-collateral pathway, regulates Schaffer-collateral long-term potentiation in the hippocampus and is required for memory acquisition and synaptic plasticity. Involved in skin desquamation and keratinocyte proliferation. Plays a role in the secondary phase of pathogenesis following spinal cord injury.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KLK8
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR