Anti-LTF Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-5810R-CY5.5
10423-458EA 810.86 USD
10423-458
Anti-LTF Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Transferrins are iron binding transport proteins which can bind two Fe(3+) ions in association with the binding of an anion, usually bicarbonate.Lactotransferrin has antimicrobial activity which depends on the extracellular cation concentration.Lactoferroxins A, B and C have opioid antagonist activity. Lactoferroxin A shows preference for mu-receptors, while lactoferroxin B and C have somewhat higher degrees of preference for kappa-receptors than for mu-receptors.The lactotransferrin transferrin-like domain 1 functions as a serine protease of the peptidase S60 family that cuts arginine rich regions. This function contributes to the antimicrobial activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LTF
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR