Anti-LGALS7 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5809R-CY3
10424-062EA 811.1 USD
10424-062
Anti-LGALS7 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Could be involved in cell-cell and/or cell-matrix interactions necessary for normal growth control. Pro-apoptotic protein that functions intracellularly upstream of JNK activation and cytochrome c release.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LGALS7
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR