Anti-NAIP Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5804R-CY3
10423-958EA 811.1 USD
10423-958
Anti-NAIP Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Prevents motor-neuron apoptosis induced by a variety of signals. Possible role in the prevention of spinal muscular atrophy that seems to be caused by inappropriate persistence of motor-neuron apoptosis: mutated or deleted forms of NAIP have been found in individuals with severe spinal muscular atrophy.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NAIP
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR