Anti-BIRC3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-5803R-A647
10423-930EA 811.1 USD
10423-930
Anti-BIRC3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Apoptotic suppressor. The BIR motifs region interacts with TNF receptor associated factors 1 and 2 (TRAF1 and TRAF2) to form an heteromeric complex, which is then recruited to the tumor necrosis factor receptor 2 (TNFR2).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BIRC3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR