Anti-BAG3 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5799R-CY7
10423-868EA 811.1 USD
10423-868
Anti-BAG3 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Inhibits the chaperone activity of HSP70/HSC70 by promoting substrate release. Has anti-apoptotic activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BAG3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR