Anti-BCL2L12 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-5797R-A555
10423-792EA 811.1 USD
10423-792
Anti-BCL2L12 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Expressed mainly in breast, thymus, prostate, fetal liver, colon, placenta, pancreas, small intestine, spinal cord, kidney, and bone marrow and to a lesser extent in many other tissues. Isoform 2 is primarily expressed in skeletal muscle.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L12
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR