Anti-BCL2L10 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5796R-CY3
10423-774EA 811.1 USD
10423-774
Anti-BCL2L10 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Promotes cell survival. Suppresses apoptosis induced by BAX but not BAK.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR