Anti-BCL2L10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5796R-A350
10423-764EA 811.1 USD
10423-764
Anti-BCL2L10 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Promotes cell survival. Suppresses apoptosis induced by BAX but not BAK.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BCL2L10
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR