Anti-ROBO4 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-5795R-CY7
10423-756EA 810.86 USD
10423-756
Anti-ROBO4 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR