Anti-ROBO4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-5795R-A555
10423-744EA 811.1 USD
10423-744
Anti-ROBO4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Receptor for Slit proteins, at least for SLIT2, and seems to be involved in angiogenesis and vascular patterning. May mediate the inhibition of primary endothelial cell migration by Slit proteins (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ROBO4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR