Anti-LOXL4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss

BS-5790R-A350
10423-628EA 811.1 USD
10423-628
Anti-LOXL4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
May modulate the formation of a collagenous extracellular matrix.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LOXL4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR