Anti-RELM beta Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5774R-CY3
10423-252EA 811.1 USD
10423-252
Anti-RELM beta Rabbit Polyclonal Antibody (Cy3®)
Antibodies
The cysteine-rich, adipose tissue-specific, secretory factor resistin (resistance to insulin, also known as ADSF) is a secreted hormone that potentially links obesity to diabetes. Resistin is rich in serine and cysteine residues and contains a unique cysteine repeat motif. Resistin and the resistin-like molecules share the characteristic cysteine composition and other signature features. Resistin-like a is a secreted protein that has restricted tissue distribution and is most highly expressed in adipose tissue. Another family member, Resistin-like b, is a secreted protein expressed only in the gastrointestinal tract, particularly in the colon, in both mouse and human. Resistin-like b expression is highest in proliferative epithelial cells and is markedly increased in tumors, suggesting a role in intestinal proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RELM beta
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR