Anti-FGF22 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-5773R-A555
10423-222EA 810.86 USD
10423-222
Anti-FGF22 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
FGF22 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The mouse homolog of this gene was found to be preferentially expressed in the inner root sheath of the hair follicle, which suggested a role in hair development.Fibroblast Growth Factor-22 stimulates the proliferation and activation of cells that express FGF receptor.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF22
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR