Anti-FGF20 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5772R-CY3
10423-204EA 811.1 USD
10423-204
Anti-FGF20 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
FGF20 is secreted heparin binding growth factor that is a member of the FGF family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion by promoting cellular proliferation and differentiation. The gene for FGF20 was shown to be expressed in normal brain, particularly the cerebellum. FGF20 signals through the FGFR 2c and 3c and is expressed during limb and brain development.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF20
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR