Anti-FGF17 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5770R-CY3
10423-156EA 811.1 USD
10423-156
Anti-FGF17 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
FGF17 belongs to the heparin-binding growth factors family. It may be a signaling molecule in the induction and patterning of the embryonic brain and is preferentially expressed in the embryonic brain. There are two named isoforms.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF17
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR