Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-FGF16 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
BS-5769R-CY5.5
10423-136
Anti-FGF16 Rabbit Polyclonal Antibody (Cy5.5®)
FGF16 belongs to the heparin-binding growth factors family. It induces hepatocellular proliferation, but has no biological effect on the heart.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF16
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat

Order Now


 

ORDER

SPECIFICATIONS

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty