Anti-FGF16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-5769R-A555
10423-126EA 811.1 USD
10423-126
Anti-FGF16 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
FGF16 belongs to the heparin-binding growth factors family. It induces hepatocellular proliferation, but has no biological effect on the heart.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: FGF16
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR