Anti-PRKD1 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss

BS-5562R-CY3
10417-646EA 811.12 USD
10417-646
Anti-PRKD1 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
Members of the protein kinase C (PKC) family function in many extracellular receptor-mediated signal transduction pathways. See PRKCA (MIM 176960) for further background information. The PRKCM gene encodes a cytosolic serine-threonine kinase that binds to the trans-Golgi network and regulates the fission of transport carriers specifically destined to the cell surface.[supplied by OMIM]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRKD1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR