Anti-MAPK9 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss

BS-5487R-CY5
10416-774EA 811.12 USD
10416-774
Anti-MAPK9 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
phosphorylated at the Thr-Pro-Tyr phosphorylation motif instead of the characteristic MAP kinase Thr-Glu-Tyr motif. JNK2 (p54a, SAPK1a), along with JNK1 and JNK3, is thought to play an important role in nuclear signal transduction through its environmental stress activation and subsequent phosphorylation of the nuclear transcription factor p53.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAPK9
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR