Anti-MEF2C Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
BS-5479R-A647
10418-424EA 746.43 USD
10418-424
Anti-MEF2C Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
MEF2C is a transcription activator which binds specifically to the MEF2 element present in the regulatory regions of many muscle-specific genes. This protein controls cardiac morphogenesis and myogenesis, and is also involved in vascular development. It may also be involved in neurogenesis and in the development of cortical architecture.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MEF2C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR