Anti-FLNA Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
BS-5351R-CY5
10413-894EA 746.43 USD
10413-894
Anti-FLNA Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Promotes orthogonal branching of actin filaments and links actin filaments to membrane glycoproteins. Anchors various transmembrane proteins to the actin cytoskeleton and serves as a scaffold for a wide range of cytoplasmic signaling proteins. Interaction with FLNA may allow neuroblast migration from the ventricular zone into the cortical plate. Tethers cell surface-localized furin, modulates its rate of internalization and directs its intracellular trafficking.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Filamin A
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR