Anti-EP300 Rabbit Polyclonal Antibody (Cy3®)

Supplier: Bioss
BS-5341R-CY3
10413-660EA 811.12 USD
10413-660
Anti-EP300 Rabbit Polyclonal Antibody (Cy3®)
Antibodies
KAT3B/p300 is a transcriptional adapter protein (300 kDa)which is characterized by three cysteine and histidine rich regions and its C-terminus specifically binds the adenovirus E1A protein. KAT3B and associated proteins are components of TATA-binding protein (TBP)complexes. Protein kinase A mediated CREB phosphorylation results in the binding of CREB to a 265 kDa nuclear protein designated KAT3A/CBP(for CREB-binding protein). KAT3B and KAT3A are homologous to each other.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EP300
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR