Anti-RBL2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Supplier: Bioss

BS-4984R-HRP
10405-290EA 811.1 USD
10405-290
Anti-RBL2 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
p130 is related both in structure and function to the retinoblastoma tumor suppressor protein and p107 (collectively known as pocket proteins) and is known to regulate the activity of E2F transcription factors. E2F transcription factors regulate the expression of a number of genes important in cell proliferation, particularly those involved in the progression through G1 and into the S phase of the cell cycle. Binding of p130 converts E2F transcription factors from transcriptional activators to transcriptional repressors.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: RBL2
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR