Anti-RBL2 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss

BS-4984R-CY5.5
10401-138EA 811.1 USD
10401-138
Anti-RBL2 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
p130 is related both in structure and function to the retinoblastoma tumor suppressor protein and p107 (collectively known as pocket proteins) and is known to regulate the activity of E2F transcription factors. E2F transcription factors regulate the expression of a number of genes important in cell proliferation, particularly those involved in the progression through G1 and into the S phase of the cell cycle. Binding of p130 converts E2F transcription factors from transcriptional activators to transcriptional repressors.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RBL2
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR