Anti-LAMA1 Rabbit Polyclonal Antibody (Cy3®)

BS-4973R-CY3
10401-100EA 382.17 USD
10401-100
Anti-LAMA1 Rabbit Polyclonal Antibody (Cy3®)
Binding to cells via a high affinity receptor, laminin is thought to mediate the attachment, migration and organization of cells into tissues during embryonic development by interacting with other extracellular matrix components.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LAMA1
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR