Anti-PCK1 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss

BS-4972R-CY7
10399-788EA 811.1 USD
10399-788
Anti-PCK1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
PCK1 is a main control point for the regulation of gluconeogenesis. This cytosolic enzyme, along with GTP, catalyzes the formation of phosphoenolpyruvate from oxaloacetate, with the release of carbon dioxide and GDP. The expression of the corresponding gene can be regulated by insulin, glucocorticoids, glucagon, cAMP, and diet. A mitochondrial isozyme has also been characterized.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCK1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR