Anti-PAPOG Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss

BS-4968R-A488
10406-060EA 810.85 USD
10406-060
Anti-PAPOG Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
Responsible for the post-transcriptional adenylation of the 3'-terminal of mRNA precursors and several small RNAs including signal recognition particle (SRP) RNA, nuclear 7SK RNA, U2 small nuclear RNA, and ribosomal 5S RNA.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAPOG
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR