Anti-PAPOA Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Supplier: Bioss

BS-4966R-A555
10406-038EA 811.1 USD
10406-038
Anti-PAPOA Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Polymerase that creates the 3'-poly(A) tail of mRNA's. Also required for the endoribonucleolytic cleavage reaction at some polyadenylation sites. May acquire specificity through interaction with a cleavage and polyadenylation specificity factor (CPSF) at its C-terminus.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAPOA
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR