Anti-MT3 Rabbit Polyclonal Antibody (Biotin)

BS-4964R-BIO
10399-756EA 529.29 USD
10399-756
Anti-MT3 Rabbit Polyclonal Antibody (Biotin)
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.

Recommended Dilutions: ELISA(1:500-1000; IHC-P: 1:100-500

Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR