Anti-MT3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss

BS-4964R-A647
10399-754EA 811.1 USD
10399-754
Anti-MT3 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Binds heavy metals. Contains three zinc and three copper atoms per polypeptide chain and only a negligible amount of cadmium. Inhibits survival and neurite formation of cortical neurons in vitro.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MT3
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR